Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142238.1 | NP_001135710.1 | 2 - 3 | 389 | 97 | |
XM_012968126.1 | XP_012823580.1 | 3 - 4 | 97 | |||
GenBank mRNA | BC168836.1 | - | 2 - 3 | 389 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |