Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142826.2 | NP_001136298.1 | 1 - 2 | 95 | 60 | |
XM_012970193.2 | XP_012825647.1 | 1 - 2 | 115 | 60 | ||
XM_012970194.2 | XP_012825648.1 | 1 - 2 | 115 | 60 | ||
XM_012970195.2 | XP_012825649.1 | 1 - 2 | 115 | 60 | ||
XM_012970196.2 | XP_012825650.1 | 1 - 2 | 115 | 60 | ||
XM_012970197.2 | XP_012825651.1 | 1 - 2 | 115 | 60 | ||
GenBank mRNA | CU075851.1 | - | 1 - 2 | 248 | 60 | |
EU220008.1 | - | 1 - 2 | 97 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |