Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142909.1 | NP_001136381.1 | 1 - 2 | 372 | 75 | |
GenBank mRNA | BC161420.1 | - | 1 - 2 | 372 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |