Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143923.1 | NP_001137395.1 | 3 - 4 | 1123 | 99 | |
XM_012952542.2 | XP_012807996.1 | 3 - 4 | 1762 | 99 | ||
XM_012952543.2 | XP_012807997.1 | 3 - 4 | 1566 | 99 | ||
GenBank mRNA | BC121572.1 | - | 2 - 3 | 418 | 99 | |
BC161272.1 | - | 3 - 4 | 794 | 99 | ||
EU422980.1 | - | 3 - 4 | 1123 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |