Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143925.1 | NP_001137397.1 | 4 - 5 | 418 | 102 | |
XM_018092949.1 | XP_017948438.1 | 4 - 5 | 601 | 102 | ||
GenBank mRNA | CR761197.2 | - | 4 - 5 | 513 | 102 | |
EU422982.1 | - | 4 - 5 | 418 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |