Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137102.1 | NP_001130574.1 | 1142 | 79 | ||
GenBank mRNA | BT033842.1 | - | 1142 | 79 | ||
BT042981.1 | - | 1157 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |