Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137541.1 | NP_001131013.1 | 1577 | 59 | ||
GenBank mRNA | BT063342.1 | - | 1598 | 59 | ||
BT064177.1 | - | 1675 | 59 | |||
BT083882.1 | - | 1598 | 59 | |||
FM211452.1 | - | 1573 | 59 | |||
FM212634.1 | - | 1577 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |