Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: his1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111960.1 | NP_001105430.1 | 186 | 84 | ||
GenBank mRNA | X57077.1 | - | 186 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Zea mays |