Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112199.1 | NP_001105669.1 | 1142 | 69 | ||
GenBank mRNA | AY264369.1 | - | 1142 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |