Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112585.1 | NP_001106055.1 | 1351 | 69 | ||
GenBank mRNA | BT041100.1 | - | 1264 | 69 | ||
BT064625.1 | - | 1330 | 69 | |||
EF650835.1 | - | 1351 | 69 | |||
EU971053.1 | - | 1293 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Zea mays |