Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100286177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159065.1 | NP_001152537.1 | 715 | 66 | ||
GenBank mRNA | BT062201.1 | - | 695 | 66 | ||
EU975999.1 | - | 715 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |