Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111724.1 | NP_001105194.1 | 1809 | 65 | ||
GenBank mRNA | AY059416.1 | - | 1652 | 65 | ||
AY124703.1 | - | 1809 | 65 |