Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pep7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112033.1 | NP_001105503.1 | 740 | 78 | ||
GenBank mRNA | AB012228.1 | - | 740 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |