Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112247.1 | NP_001105717.1 | 1209 | 66 | ||
GenBank mRNA | Z38111.1 | - | 1209 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |