Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112425.1 | NP_001105895.1 | 2753 | 69 | ||
GenBank mRNA | U85495.1 | - | 2753 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Zea mays |