Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112519.1 | NP_001105989.1 | 1646 | 77 | ||
GenBank mRNA | BT066397.1 | - | 1630 | 77 | ||
DQ832257.1 | - | 1646 | 77 |