Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111699.1 | NP_001105169.1 | 1193 | 84 | ||
GenBank mRNA | AY029315.1 | - | 1193 | 84 | ||
AY103929.1 | - | 1195 | 84 | |||
BT035073.1 | - | 1458 | 84 | |||
BT066636.1 | - | 1202 | 84 |