Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111892.1 | NP_001105362.1 | 1150 | 93 | ||
GenBank mRNA | AY111119.1 | - | 1150 | 93 | ||
EU961361.1 | - | 1271 | 93 | |||
U10076.1 | - | 1150 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Zea mays |