Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138792.2 | NP_001132264.2 | 1457 | 69 | ||
NM_001204251.1 | NP_001191180.1 | 1331 | 69 | |||
GenBank mRNA | BT036036.1 | - | 788 | 69 | ||
BT064122.1 | - | 1603 | 69 | |||
FJ750248.1 | - | 1457 | 69 | |||
FJ750249.1 | - | 1331 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Zea mays |