Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101389.3 | NP_172973.1 | 8 - 9 | 3715 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |