Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332166.1 | NP_001319010.1 | 1 - 2 | 243 | 143 | |
NM_001332167.1 | NP_001322448.1 | 2 - 3 | 254 | 143 | ||
NM_101415.4 | NP_172999.1 | 2 - 3 | 253 | 143 | ||
GenBank mRNA | AY069887.1 | - | 1 - 2 | 168 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |