Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035973.3 | NP_001031050.1 | 13 - 14 | 2572 | 73 | |
NM_001035974.4 | NP_001031051.1 | 14 - 15 | 2306 | 73 | ||
NM_101443.4 | NP_563981.1 | 14 - 15 | 2466 | 73 | ||
NM_179341.3 | NP_849672.1 | 15 - 16 | 2376 | 73 | ||
GenBank mRNA | AY046013.1 | - | 14 - 15 | 2236 | 73 | |
AY142619.1 | - | 13 - 14 | 2072 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |