Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101464.4 | NP_173048.3 | 12 - 13 | 2299 | 151 | |
GenBank mRNA | AJ291831.1 | - | 12 - 13 | 1260 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |