Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101466.3 | NP_563986.1 | 3 - 4 | 938 | 96 | |
GenBank mRNA | AY057703.1 | - | 3 - 4 | 785 | 96 | |
AY084966.1 | - | 3 - 4 | 785 | 96 | ||
AY124875.1 | - | 3 - 4 | 769 | 96 | ||
KJ139005.1 | - | 3 - 4 | 769 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |