Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101475.3 | NP_173059.1 | 7 - 8 | 1252 | 84 | |
GenBank mRNA | AF487269.1 | - | 7 - 8 | 1051 | 84 | |
BT025624.1 | - | 7 - 8 | 1051 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |