Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCB3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101499.4 | NP_173083.3 | 20 - 21 | 1604 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |