Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101587.2 | NP_173169.2 | 19 - 20 | 2588 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |