Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AlaAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101591.6 | NP_173173.3 | 4 - 5 | 829 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |