Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332318.1 | NP_001321828.1 | 8 - 9 | 945 | 89 | |
NM_101628.4 | NP_564030.2 | 8 - 9 | 830 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |