Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BETA-TIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101644.3 | NP_173223.1 | 1 - 2 | 324 | 58 | |
GenBank mRNA | AF026275.1 | - | 1 - 2 | 158 | 58 | |
AK229035.1 | - | 1 - 2 | 242 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |