Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101850.2 | NP_564100.1 | 4 - 5 | 487 | 63 | |
GenBank mRNA | AY045614.1 | - | 4 - 5 | 389 | 63 | |
BT001012.1 | - | 3 - 4 | 186 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |