Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332435.1 | NP_001319049.1 | 7 - 8 | 695 | 88 | |
NM_001332436.1 | NP_001322430.1 | 7 - 8 | 697 | 88 | ||
NM_001332437.1 | NP_001322431.1 | 7 - 8 | 690 | 88 | ||
NM_001332438.1 | NP_001322429.1 | 7 - 8 | 695 | 88 | ||
NM_101862.5 | NP_173436.4 | 6 - 7 | 714 | 88 | ||
GenBank mRNA | FM213366.1 | - | 6 - 7 | 709 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |