Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103786.3 | NP_175323.1 | 4 - 5 | 1055 | 152 | |
GenBank mRNA | BT002935.1 | - | 4 - 5 | 948 | 152 | |
BT005638.1 | - | 4 - 5 | 874 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |