Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345737.1 | NP_001332639.1 | 11 - 12 | 1692 | 98 | |
NM_001345738.1 | NP_001332640.1 | 11 - 12 | 1667 | 98 | ||
NM_126000.2 | NP_569025.2 | 10 - 11 | 1397 | 98 | ||
NM_180946.3 | NP_851277.1 | 11 - 12 | 1727 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |