Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345737.1 | NP_001332639.1 | 13 - 14 | 1930 | 92 | |
NM_001345738.1 | NP_001332640.1 | 13 - 14 | 1905 | 92 | ||
NM_126000.2 | NP_569025.2 | 12 - 13 | 1635 | 92 | ||
NM_180946.3 | NP_851277.1 | 13 - 14 | 1965 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |