Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345810.1 | NP_001332428.1 | 2 | 901 | 141 | |
NM_126106.3 | NP_201507.1 | 1 | 907 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |