Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126111.4 | NP_201512.1 | 4 - 5 | 534 | 121 | |
GenBank mRNA | BT028946.1 | - | 4 - 5 | 441 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |