Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126173.4 | NP_565252.1 | 3 - 4 | 984 | 73 | |
GenBank mRNA | AB054096.1 | - | 3 - 4 | 814 | 73 | |
AF145045.1 | - | 3 - 4 | 814 | 73 | ||
AF360172.1 | - | 3 - 4 | 834 | 73 | ||
AY056347.1 | - | 3 - 4 | 814 | 73 | ||
AY084784.1 | - | 3 - 4 | 955 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |