Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126218.3 | NP_178266.1 | 1 | 2072 | 89 | |
GenBank mRNA | AY054160.1 | - | 1 | 2037 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |