Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF73-II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335074.1 | NP_001325256.1 | 11 - 12 | 1398 | 66 | |
NM_126234.3 | NP_178282.2 | 12 - 13 | 1312 | 66 | ||
GenBank mRNA | AK221561.1 | - | 12 - 13 | 1244 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |