Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126278.3 | NP_027422.1 | 10 - 11 | 1106 | 109 | |
GenBank mRNA | AB036427.1 | - | 10 - 11 | 844 | 109 | |
AK117222.1 | - | 10 - 11 | 873 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |