Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126295.1 | NP_178343.1 | 4 - 5 | 275 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |