Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111003.5 | NP_186786.2 | 4 - 5 | 1666 | 93 | |
GenBank mRNA | AY136347.1 | - | 4 - 5 | 1666 | 93 | |
BT010594.1 | - | 4 - 5 | 1644 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |