Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111003.5 | NP_186786.2 | 6 - 7 | 2116 | 80 | |
GenBank mRNA | AY136347.1 | - | 6 - 7 | 2116 | 80 | |
BT010594.1 | - | 6 - 7 | 2094 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |