Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111003.5 | NP_186786.2 | 8 - 9 | 2237 | 149 | |
GenBank mRNA | AY136347.1 | - | 8 - 9 | 2237 | 149 | |
BT010594.1 | - | 8 - 9 | 2215 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |