Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084626.2 | NP_001078095.1 | 2 - 3 | 622 | 124 | |
NM_001084627.1 | NP_001078096.1 | 2 - 3 | 335 | 124 | ||
NM_001084628.2 | NP_001078097.1 | 3 - 4 | 959 | 124 | ||
NM_001084629.2 | NP_001078098.1 | 2 - 3 | 447 | 124 | ||
NM_001337419.1 | NP_001327801.1 | 3 - 4 | 1440 | 124 | ||
NM_001337420.1 | NP_001327802.1 | 1 - 2 | 1949 | 124 | ||
NM_001337421.1 | NP_001319453.1 | 3 - 4 | 526 | 124 | ||
NM_001337422.1 | NP_001327800.1 | 2 - 3 | 885 | 124 | ||
NM_001337423.1 | NP_001327797.1 | 3 - 4 | 765 | 124 | ||
NM_001337424.1 | NP_001327798.1 | 2 - 3 | 1841 | 124 | ||
NM_001337425.1 | NP_001327799.1 | 2 - 3 | 1560 | 124 | ||
NM_111128.2 | NP_566177.1 | 3 - 4 | 715 | 124 | ||
GenBank mRNA | AY084915.1 | - | 3 - 4 | 548 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |