Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111136.3 | NP_186917.2 | 10 - 11 | 1485 | 103 | |
GenBank mRNA | DQ167217.1 | - | 10 - 11 | 1415 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |