Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337439.1 | NP_001325525.1 | 2 - 3 | 652 | 69 | |
NM_001337440.1 | NP_001325526.1 | 2 - 3 | 652 | 69 | ||
NM_111156.3 | NP_186937.1 | 2 - 3 | 652 | 69 | ||
GenBank mRNA | AY054640.1 | - | 2 - 3 | 563 | 69 | |
AY081499.1 | - | 2 - 3 | 483 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |