Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084941.2 | NP_001078410.1 | 1 - 2 | 490 | 84 | |
NM_001341395.1 | NP_001329642.1 | 2 - 3 | 372 | 84 | ||
NM_001341396.1 | NP_001329640.1 | 3 - 4 | 362 | 84 | ||
NM_001341397.1 | NP_001329641.1 | 3 - 4 | 337 | 84 | ||
NM_118124.3 | NP_193738.1 | 2 - 3 | 554 | 84 | ||
GenBank mRNA | AY268942.1 | - | 2 - 3 | 554 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |