Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341400.1 | NP_001327970.1 | 9 - 10 | 1140 | 149 | |
NM_001341401.1 | NP_001327969.1 | 7 - 8 | 820 | 149 | ||
NM_118132.5 | NP_193746.3 | 10 - 11 | 1233 | 149 | ||
GenBank mRNA | AK230206.1 | - | 10 - 11 | 1155 | 149 | |
BT004184.1 | - | 10 - 11 | 1147 | 149 | ||
BT005475.1 | - | 10 - 11 | 1136 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |